Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Thermo Scientific Chemicals
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
For column chromatography.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number ACR240360010
Price (MYR)
950.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
950.00
Add to cart
Chemical Identifiers
CAS120-57-0
IUPAC Name2H-1,3-benzodioxole-5-carbaldehyde
Molecular FormulaC8H6O3
InChI KeySATCULPHIDQDRE-UHFFFAOYSA-N
SMILESO=CC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Melting point35°C to 38°C
Appearance (Color)White to light brown
Infrared spectrumConforms
GC>=98.5 %
For column chromatography
RUO – Research Use Only