Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
CAS: 25013-16-5 | C11H16O2 | 180.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR235235000
Price (MYR)
1,068.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,068.00
Add to cart
Specifications
Chemical Name or MaterialButylated hydroxyanisole
Name Note96%
CAS25013-16-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.