Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
CAS: 25013-16-5 | C11H16O2 | 180.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR235231000
Price (MYR)
464.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
464.00
Add to cart
Chemical Identifiers
CAS59-14-3
IUPAC Name5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11BrN2O5
InChI KeyWOVKYSAHUYNSMH-RRKCRQDMSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Related substances=<0.5 % (HPLC) 5-Bromouracil
Related substances=<0.2 % (HPLC) Each unknown impurity
Related substances=<1.0 % (HPLC) Total impurities
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals