Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
CAS: 9005-64-5 | C26H50O10
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 mL
250 mL
1 L
Packaging Type:
View all
Plastic drum
Catalog number ACR233360250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Packaging Type:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or MaterialPolysorbate 20
CAS9005-64-5
ColorColorless to Yellow
Density1.1000 g/mL
Molecular FormulaC26H50O10