Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
CAS: 9005-64-5 | C26H50O10
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
25 L
Packaging Type:
View all
Catalog number ACR233360051
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
SolubilitySolubility in water: 100g/L. Other solubilities: 100g/L isopropylalcohol, xylene, insoluble in mineral oil, 10g/L propylene glycol