Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
CAS: 9005-64-5 | C26H50O10
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
5 L
25 L
Packaging Type:
View all
Glass Bottle
Catalog number ACR233360050
Price (MYR)
381.00
Add to cart
Quantity:
5 mL
Packaging Type:
Glass Bottle
Request bulk or custom format
Price (MYR)
381.00
Add to cart
Chemical Identifiers
CAS40018-25-5
IUPAC Name3-(2-chlorophenyl)-3-oxopropanenitrile
Molecular FormulaC9H6ClNO
InChI KeySBSWHTFHLWSSQS-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to orange to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%