Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
CAS: 9005-64-5 | C26H50O10
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
250 mL
5 L
25 L
Packaging Type:
View all
Glass Bottle
Catalog number ACR233360010
Price (MYR)
719.00
Add to cart
Quantity:
1 L
Packaging Type:
Glass Bottle
Request bulk or custom format
Price (MYR)
719.00
Add to cart
Specifications
Chemical Name or MaterialPolysorbate 20
CAS9005-64-5
ColorColorless to Yellow
Density1.1000 g/mL
Molecular FormulaC26H50O10