Iron(II) sulfide, fused sticks
Iron(II) sulfide, fused sticks
Iron(II) sulfide, fused sticks
Iron(II) sulfide, fused sticks
Thermo Scientific Chemicals
Iron(II) sulfide, fused sticks
CAS: 1317-37-9 | FeS | 87.91 g/mol
Have Questions?
Catalog number ACR233300010
Price (MYR)
712.00
Add to cart
Request bulk or custom format
Price (MYR)
712.00
Add to cart
Chemical Identifiers
CAS33630-99-8
IUPAC Name3-amino-1,2-dihydropyridin-2-one
Molecular FormulaC5H6N2O
InChI KeyVTSFNCCQCOEPKF-UHFFFAOYSA-N
SMILESNC1=CC=CNC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%