Petroleum ether, extra pure, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 100-140°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 100-140°C, Thermo Scientific Chemicals
CAS: 64742-49-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number ACR233020025
Price (MYR)
800.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
800.00
Add to cart
Chemical Identifiers
CAS15170-57-7
IUPAC Nameplatinum(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14O4Pt
InChI KeyKLFRPGNCEJNEKU-FDGPNNRMSA-L
SMILES[Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pt 48.0% min