Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals
Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals
Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals
Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals
CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR232890010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHexamethylenetetramine
Name Note98.5%
CAS100-97-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Flammable solid.