1-Hydroxypyrene, 99+%, Thermo Scientific Chemicals
1-Hydroxypyrene, 99+%, Thermo Scientific Chemicals
1-Hydroxypyrene, 99+%, Thermo Scientific Chemicals
1-Hydroxypyrene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hydroxypyrene, 99+%, Thermo Scientific Chemicals
CAS: 5315-79-7 | C16H10O | 218.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
Packaging:
Glass bottle
Catalog number ACR232270500
Price (MYR)
698.00
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
698.00
Add to cart
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.05% max.
Particle Size-10 Mesh