4-(Aminomethyl)benzoic acid, 99%, Thermo Scientific Chemicals
4-(Aminomethyl)benzoic acid, 99%, Thermo Scientific Chemicals
4-(Aminomethyl)benzoic acid, 99%, Thermo Scientific Chemicals
4-(Aminomethyl)benzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Aminomethyl)benzoic acid, 99%, Thermo Scientific Chemicals
CAS: 56-91-7 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR232220250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-(Aminomethyl)benzoic acid
Name Note99%
CAS56-91-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning