Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
Packaging:
View all
Catalog number ACR232110025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7791-13-1
IUPAC Nameλ²-cobalt(2+) hexahydrate dichloride
Molecular FormulaCl2CoH12O6
InChI KeyGFHNAMRJFCEERV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to purple
Titration Complexometric98.0 to 102.0 %
Calcium (Ca)=<50 ppm
Magnesium (Mg)=<20 ppm
Insoluble matter=<0.03 % (in water)
View more
For analysis
RUO – Research Use Only