n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR232100025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS3710-84-7
IUPAC NameN,N-diethylhydroxylamine
Molecular FormulaC4H11NO
InChI KeyFVCOIAYSJZGECG-UHFFFAOYSA-N
SMILESCCN(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<100 APHA
GC>=96.0 %
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
Appearance (Form)Clear liquid
Infrared spectrumConforms
View more
For HPLC
RUO – Research Use Only