n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR232100010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS404-71-7
IUPAC Name1-fluoro-3-isocyanatobenzene
Molecular FormulaC7H4FNO
InChI KeyRIKWVZGZRYDACA-UHFFFAOYSA-N
SMILESFC1=CC=CC(=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormFlakes or clear liquid as melt
Appearance (Color)Colorless to white to pale yellow
Assay (GC)≥97.0%
Identification (FTIR)Conforms
Refractive Index1.5115-1.5165 @ 20?C
For HPLC
RUO – Research Use Only