Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR232090025
Price (MYR)
999.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
999.00
Add to cart
Chemical Identifiers
CAS91-17-8
IUPAC Namedecahydronaphthalene
Molecular FormulaC10H18
InChI KeyNNBZCPXTIHJBJL-UHFFFAOYSA-N
SMILESC1CCC2CCCCC2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4720-1.4760 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
For analysis
RUO – Research Use Only