Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, +99%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR232090010
Price (MYR)
642.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
642.00
Add to cart
Chemical Identifiers
CAS698-80-6
IUPAC Name4-(chloromethyl)-1,2-difluorobenzene
Molecular FormulaC7H5ClF2
InChI KeyVTGRVYJPIVZZGS-UHFFFAOYSA-N
SMILESFC1=CC=C(CCl)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4920 to 1.4940 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
For analysis
RUO – Research Use Only