D(+)-Biotin, 98%
D(+)-Biotin, 98%
D(+)-Biotin, 98%
Thermo Scientific Chemicals
D(+)-Biotin, 98%
CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
500 mg
5 g
25 g
Packaging:
View all
Glass Bottle
Catalog number ACR230090010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS16940-81-1
IUPAC Namehydrogen hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6HP
InChI KeyLJQLCJWAZJINEB-UHFFFAOYSA-O
SMILES[H+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay Titrimetry>=55.0 %
Appearance (Color)Colorless to yellow-green
Appearance (Form)Solution