γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
CAS: 91464-90-3 | C48H80O40 | 1297.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 mg
1 g
Packaging:
Glass bottle
Catalog number ACR229900050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS84625-61-6
IUPAC Name1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one
Molecular FormulaC35H38Cl2N8O4
InChI KeyVHVPQPYKVGDNFY-ZPGVKDDISA-N
SMILESCCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point165°C to 169°C
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.5 %
View more
Chelation/Complexation reagent
RUO – Research Use Only