γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
CAS: 91464-90-3 | C48H80O40 | 1297.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
500 mg
5 g
Packaging:
Glass bottle
Catalog number ACR229900010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS546-89-4
IUPAC Namelithium(1+) acetate
Molecular FormulaC2H3LiO2
InChI KeyXIXADJRWDQXREU-UHFFFAOYSA-M
SMILES[Li+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Melting point280°C to 285°C
View more
Chelation/Complexation reagent
RUO – Research Use Only