3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
CAS: 54827-17-7 | C16H20N2 | 240.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR229280250
Price (MYR)
3,134.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,134.00
Add to cart
Specifications
Chemical Name or Material3,3′,5,5′-Tetramethylbenzidine
CAS54827-17-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen