3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
CAS: 54827-17-7 | C16H20N2 | 240.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR229280010
Price (MYR)
494.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
494.00
Add to cart
Chemical Identifiers
CAS100-97-0
IUPAC Name1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
Molecular FormulaC6H12N4
InChI KeyVKYKSIONXSXAKP-UHFFFAOYSA-N
SMILESC1N2CN3CN1CN(C2)C3
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥99.0 to ≤101.0%
Water Content (Karl Fischer Titration)≤1%
FormCrystals or powder or crystalline powder or chunks or fused solid