Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
CAS: 60-27-5 | C4H7N3O | 113.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
25 g
250 g
Packaging:
Glass bottle
Catalog number ACR228940500
Price (MYR)
690.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
690.00
Add to cart
Chemical Identifiers
CAS50-99-7
IUPAC Name6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILESOCC1OC(O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<5 ppm
Appearance (Form)Crystalline powder
Loss on drying=<0.2 % (105°C, 6 h)
Residue after ignition=<0.02 %
Iron (Fe)=<5 ppm