Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
CAS: 60-27-5 | C4H7N3O | 113.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle
Catalog number ACR228940250
Price (MYR)
511.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
511.00
Add to cart
Specifications
Chemical Name or MaterialCreatinine
CAS60-27-5
Melting Point255.0°C
ColorWhite
EINECS Number200-466-7