Pyridoxal 5-phosphate monohydrate, 99%
Pyridoxal 5-phosphate monohydrate, 99%
Pyridoxal 5-phosphate monohydrate, 99%
Pyridoxal 5-phosphate monohydrate, 99%
Thermo Scientific Chemicals
Pyridoxal 5-phosphate monohydrate, 99%
CAS: 41468-25-1 | C8H10NO6P | 247.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR228170250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil
Appearance (Color)Copper
Comment0.9 - 1.1mm thick
Total Metal Impurities0.01% max.