Carbonyl cyanide 3-chlorophenylhydrazone, 98%, Thermo Scientific Chemicals
Carbonyl cyanide 3-chlorophenylhydrazone, 98%, Thermo Scientific Chemicals
Carbonyl cyanide 3-chlorophenylhydrazone, 98%, Thermo Scientific Chemicals
Carbonyl cyanide 3-chlorophenylhydrazone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbonyl cyanide 3-chlorophenylhydrazone, 98%, Thermo Scientific Chemicals
CAS: 555-60-2 | C9H5ClN4 | 204.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number ACR228131000
Price (MYR)
486.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
486.00
Add to cart
Specifications
Chemical Name or MaterialCarbonyl cyanide 3-chlorophenylhydrazone
Name Note99+%
CAS555-60-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.