D(+)-Camphor, 97%, Thermo Scientific Chemicals
D(+)-Camphor, 97%, Thermo Scientific Chemicals
D(+)-Camphor, 97%, Thermo Scientific Chemicals
D(+)-Camphor, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D(+)-Camphor, 97%, Thermo Scientific Chemicals
CAS: 464-49-3 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR227841000
Price (MYR)
459.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
459.00
Add to cart
Specifications
Chemical Name or MaterialD(+)-Camphor
Name Note97%
CAS464-49-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.