(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
Thermo Scientific Chemicals
(+)-5-Fluoro-2'-deoxyuridine, 99+%
CAS: 50-91-9 | C9H11FN2O5 | 246.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Packaging:
Glass bottle
Catalog number ACR227605000
Price (MYR)
3,342.00
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,342.00
Add to cart
Chemical Identifiers
CAS1317-70-0
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to gray
Total Metal Impurities0.1% max.
Particle Size32nm average