(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
Thermo Scientific Chemicals
(+)-5-Fluoro-2'-deoxyuridine, 99+%
CAS: 50-91-9 | C9H11FN2O5 | 246.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number ACR227601000
Price (MYR)
1,148.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,148.00
Add to cart
Chemical Identifiers
CAS13598-36-2
IUPAC Namedihydroxy(oxo)-λ⁵-phosphanylium
Molecular FormulaH2O3P
InChI KeyXNQULTQRGBXLIA-UHFFFAOYSA-O
SMILESO[P+](O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormFlakes or crystals or crystalline powder or powder
Water Content (Karl Fischer Titration)<1.5%
Appearance (Color)White to pale cream
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%