β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
CAS: 68168-23-0 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR227281000
Price (MYR)
1,111.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,111.00
Add to cart
Specifications
Chemical Name or Materialβ-Cyclodextrin hydrate
Name Note99%
CAS68168-23-0
CAS Min %98.5
CAS Max %100.0
View more
Chelation/Complexation reagent
RUO – Research Use Only