β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
CAS: 68168-23-0 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR227280250
Price (MYR)
493.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
493.00
Add to cart
Chemical Identifiers
CAS50-99-7
IUPAC Name6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILESOCC1OC(O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation+52.6? to 53.2? (dry basis, c=10 in 0.012N NH₄OH)
FormWhite crystalline powder
Water Content (Karl Fischer Titration)≤ 1.0%
Identification (FTIR)Conforms
Chelation/Complexation reagent
RUO – Research Use Only