β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
CAS: 68168-23-0 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR227280050
Price (MYR)
339.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
339.00
Add to cart
Chemical Identifiers
CAS9010-66-6
MDL NumberMFCD00062414
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to straw yellow to tan
Infrared spectrumConforms
Loss on drying=<8.0 % (105°C, 2 h)
Appearance (Form)Powder
Residue after ignition=<2.0 % (800°C)
View more
Chelation/Complexation reagent
RUO – Research Use Only