N-Acetylneuraminic acid, 99%, Thermo Scientific Chemicals
N-Acetylneuraminic acid, 99%, Thermo Scientific Chemicals
N-Acetylneuraminic acid, 99%, Thermo Scientific Chemicals
N-Acetylneuraminic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Acetylneuraminic acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
Catalog number ACR227040250
Price (MYR)
448.00
Add to cart
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
448.00
Add to cart
Specifications
Chemical Name or MaterialN-Acetylneuraminic acid
Name Note98%
CAS131-48-6
CAS Min %98.5
Health Hazard 1HYGROSCOPIC