1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
CAS: 6004-24-6 | C21H40ClNO | 358.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number ACR226995000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS13455-21-5
IUPAC Namepotassium dihydrate fluoride
Molecular FormulaFH4KO2
InChI KeyMZFXHSQFQLKKFD-UHFFFAOYSA-M
SMILESO.O.[F-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min