1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
CAS: 6004-24-6 | C21H40ClNO | 358.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number ACR226991000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Hexadecylpyridinium chloride monohydrate
Name Note96.0-101.0%
CAS6004-24-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger