5-Aminouracil, 98%, Thermo Scientific Chemicals
5-Aminouracil, 98%, Thermo Scientific Chemicals
5-Aminouracil, 98%, Thermo Scientific Chemicals
5-Aminouracil, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Aminouracil, 98%, Thermo Scientific Chemicals
CAS: 932-52-5 | C4H5N3O2 | 127.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR226820250
Price (MYR)
1,169.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,169.00
Add to cart
Specifications
Chemical Name or Material5-Aminouracil
Name Note98%
CAS932-52-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning