Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 929-77-1 | C23H46O2 | 354.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR226810050
Price (MYR)
2,125.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
2,125.00
Add to cart
Chemical Identifiers
CAS7790-69-4
IUPAC Namelithium(1+) nitrate
Molecular FormulaLiNO3
InChI KeyIIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES[Li+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder
Titration after Ion exchange>=99.0 %
Appearance (Color)White
Loss on drying=<1 % (200°C)