Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 929-77-1 | C23H46O2 | 354.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR226810010
Price (MYR)
700.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
700.00
Add to cart
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%