Thymidine, 99+%
Thymidine, 99+%
Thymidine, 99+%
Thermo Scientific Chemicals
Thymidine, 99+%
CAS: 50-89-5 | C10H14N2O5 | 242.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR226740250
Price (MYR)
1,685.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,685.00
Add to cart
Specifications
Chemical Name or MaterialThymidine
CAS50-89-5
Absorbance0.62 to 0.68 (A250/A260),0.70 to 0.76 (A280/A260)
Health Hazard 1Warning
Melting Point187.0°C to 189.0°C