Thymidine, 99+%
Thymidine, 99+%
Thymidine, 99+%
Thermo Scientific Chemicals
Thymidine, 99+%
CAS: 50-89-5 | C10H14N2O5 | 242.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR226740050
Price (MYR)
656.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
656.00
Add to cart
Chemical Identifiers
CAS17450-56-5
IUPAC Nameethyl (2E)-4-oxo-4-phenylbut-2-enoate
Molecular FormulaC12H12O3
InChI KeyACXLBHHUHSJENU-CMDGGOBGSA-N
SMILESCCOC(=O)C=CC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5415-1.5485 @ 20?C
Appearance (Color)Clear yellow
FormLiquid
Assay (GC)≥92.0%