2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
Thermo Scientific Chemicals
2'-Deoxycytidine, 99+%
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR226720250
Price (MYR)
6,586.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,586.00
Add to cart
Chemical Identifiers
CAS85-01-8
IUPAC Namephenanthrene
Molecular FormulaC14H10
InChI KeyYNPNZTXNASCQKK-UHFFFAOYSA-N
SMILESC1=CC=C2C(C=CC3=CC=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or grey
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)97-101°C