2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
Thermo Scientific Chemicals
2'-Deoxycytidine, 99+%
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number ACR226720050
Price (MYR)
1,652.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,652.00
Add to cart
Chemical Identifiers
CAS39098-97-0
IUPAC Name2-(thiophen-2-yl)acetyl chloride
Molecular FormulaC6H5ClOS
InChI KeyAJYXPNIENRLELY-UHFFFAOYSA-N
SMILESClC(=O)CC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5500 to 1.5520 (20°C, 589 nm)
Appearance (Color)Clear brown
GC>=98.5 %