2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
Thermo Scientific Chemicals
2'-Deoxycytidine, 99+%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR226720010
Price (MYR)
582.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
582.00
Add to cart
Chemical Identifiers
CAS14347-78-5
IUPAC Name(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
Molecular FormulaC6H12O3
InChI KeyRNVYQYLELCKWAN-UHFFFAOYNA-N
SMILESCC1(C)OCC(CO)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4330 to 1.4360 (20°C, 589 nm)