Aristolochic acid I, 96%, Thermo Scientific Chemicals
Aristolochic acid I, 96%, Thermo Scientific Chemicals
Aristolochic acid I, 96%, Thermo Scientific Chemicals
Aristolochic acid I, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aristolochic acid I, 96%, Thermo Scientific Chemicals
CAS: 313-67-7 | C17H11NO7 | 341.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number ACR226091000
Price (MYR)
1,459.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,459.00
Add to cart
Chemical Identifiers
CAS1460-57-7
IUPAC Name(1R,2R)-cyclohexane-1,2-diol
Molecular FormulaC6H12O2
InChI KeyPFURGBBHAOXLIO-PHDIDXHHSA-N
SMILESO[C@@H]1CCCC[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)99.0-106.0?C
Appearance (Color)White to cream
Assay (GC)≥97.5%