Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals
Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals
Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals
Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals
CAS: 3087-36-3 | C8H20O4Ti | 228.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number ACR223762500
Price (MYR)
1,399.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,399.00
Add to cart
Chemical Identifiers
CAS7722-84-1
IUPAC Nameperoxol
Molecular FormulaH2O2
InChI KeyMHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILESOO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Heavy metals=<10 ppm
Chloride (Cl)=<2 ppm
Stabilizerabout 0.03% stabilizers
Color scale=<10 APHA
View more
Lewis acid
RUO – Research Use Only