hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
CAS: 27988-77-8 | AuCl4H | 339.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR223620050
Price (MYR)
6,991.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,991.00
Add to cart
Chemical Identifiers
CAS6099-90-7
IUPAC Namebenzene-1,3,5-triol dihydrate
Molecular FormulaC6H10O5
InChI KeyMPYXTIHPALVENR-UHFFFAOYSA-N
SMILESO.O.OC1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=98.5 %