hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
CAS: 27988-77-8 | AuCl4H | 339.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR223620010
Price (MYR)
2,146.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,146.00
Add to cart
Chemical Identifiers
CAS135-19-3
IUPAC Namenaphthalen-2-ol
Molecular FormulaC10H8O
InChI KeyJWAZRIHNYRIHIV-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)White to beige to pink to light brown
Appearance (Form)Powder or flakes
Infrared spectrumConforms
Melting point120°C to 124°C