1-Decyne, 98%, Thermo Scientific Chemicals
1-Decyne, 98%, Thermo Scientific Chemicals
1-Decyne, 98%, Thermo Scientific Chemicals
1-Decyne, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Decyne, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number ACR223430250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Decyne
Name Note98%
CAS764-93-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning