Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals
Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals
Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals
Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals
CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass bottle
Catalog number ACR223320025
Price (MYR)
1,550.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,550.00
Add to cart
Chemical Identifiers
CAS490-79-9
IUPAC Name2,5-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyWXTMDXOMEHJXQO-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point204°C to 207°C
Titration with NaOH>=98.5 %
View more
C-X bond formation reagent
RUO – Research Use Only