Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number ACR223310010
Price (MYR)
1,679.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,679.00
Add to cart
Chemical Identifiers
CAS591-78-6
IUPAC Namehexan-2-one
Molecular FormulaC6H12O
InChI KeyQQZOPKMRPOGIEB-UHFFFAOYSA-N
SMILESCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3995 to 1.4015 (20°C, 589 nm)
Oxidation reagent for analysis
RUO – Research Use Only